Gene Transcripts

Gene Symbol
PTH2
Entrez Gene ID
113091
Gene Name
parathyroid hormone 2
Gene Aliases
TIP39
Chromosome Location
Chr. 19: 49422414 - 49423441 on Build GRCh38
UniGene ID
Hs.339845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178449.3 NP_848544.1 2 329
GenBank mRNA AY048588.1 - 2 230
BC093995.1 - 2 268 7939840
BC093997.1 - 2 272 7939842

View Full Product Details