Gene Transcripts
- Gene Symbol
- RPL41
- Entrez Gene ID
- 6171
- Gene Name
- ribosomal protein L41
- Gene Aliases
- L41
- Chromosome Location
- Chr. 12: 56116590 - 56117832 on Build GRCh38
- UniGene ID
- Hs.112553
- Species
- Homo sapiens
- Gene Symbol
- RPL41P5
- Entrez Gene ID
- 6172
- Gene Name
- ribosomal protein L41 pseudogene 5
- Gene Aliases
- RPL41L
- Chromosome Location
- Chr. 12: 93083293 - 93083716 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
S64030.1 |
- |
1 |
149 |
|
View Full Product Details