Gene Transcripts
- Gene Symbol
- ERVK-6
- Entrez Gene ID
- 64006
- Gene Name
- endogenous retrovirus group K member 6
- Gene Aliases
- ERVK6, HERV-K(C7), HERV-K108, K-Rev, c-orf, cORF
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
- Gene Symbol
- C11orf72
- Entrez Gene ID
- 100505621
- Gene Name
- chromosome 11 open reading frame 72
- Gene Aliases
- Chromosome Location
- Chr. 11: 67602880 - 67606706 on Build GRCh38
- UniGene ID
- Hs.382074
- Species
- Homo sapiens
- Gene Symbol
- CFLAR
- Entrez Gene ID
- 8837
- Gene Name
- CASP8 and FADD like apoptosis regulator
- Gene Aliases
- CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS
- Chromosome Location
- Chr. 2: 201116015 - 201172688 on Build GRCh38
- UniGene ID
- Hs.390736
- Species
- Homo sapiens
- Gene Symbol
- DHTKD1
- Entrez Gene ID
- 55526
- Gene Name
- dehydrogenase E1 and transketolase domain containing 1
- Gene Aliases
- AMOXAD, CMT2Q
- Chromosome Location
- Chr. 10: 12068917 - 12123228 on Build GRCh38
- UniGene ID
- Hs.104980
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL359587.2 |
- |
4 |
3301 |
|
- Gene Symbol
- ERVK-19
- Entrez Gene ID
- 100862685
- Gene Name
- endogenous retrovirus group K member 19
- Gene Aliases
- HERV-K(C19), P1.8, PR, Protease, Proteinase, envK3, polymerase
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY395519.1 |
- |
2 |
419 |
|
- Gene Symbol
- ERVK-21
- Entrez Gene ID
- 100616103
- Gene Name
- endogenous retrovirus group K member 21
- Gene Aliases
- Endo-Rev2, PR, Protease, Proteinase, c12_A, envK1
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
FN806845.1 |
- |
1 |
423 |
|
- Gene Symbol
- ERVK-25
- Entrez Gene ID
- 100862683
- Gene Name
- endogenous retrovirus group K member 25
- Gene Aliases
- PR, Protease, Proteinase, c11_A
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CF227268.1 |
- |
3 |
632 |
|
- Gene Symbol
- ERVK-4
- Entrez Gene ID
- 60359
- Gene Name
- endogenous retrovirus group K member 4
- Gene Aliases
- ERVK4, HERV-K(I), HERV-KI
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
- Gene Symbol
- ERVK-9
- Entrez Gene ID
- 100616102
- Gene Name
- endogenous retrovirus group K member 9
- Gene Aliases
- PR, Protease, Proteinase, polymerase
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY371030.1 |
- |
2 |
419 |
|
- Gene Symbol
- LOC105373738
- Entrez Gene ID
- 105373738
- Gene Name
- uncharacterized LOC105373738
- Gene Aliases
- Chromosome Location
- Chr. 2: 171771784 - 171793618 on Build GRCh38
- UniGene ID
- Hs.636887
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK024142.1 |
- |
2 |
1413 |
|
- Gene Symbol
- LOC105376177
- Entrez Gene ID
- 105376177
- Gene Name
- uncharacterized LOC105376177
- Gene Aliases
- Chromosome Location
- Chr. 9: 100353036 - 100390489 on Build GRCh38
- UniGene ID
- Hs.556827
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056351.1 |
- |
4 |
1962 |
|
- Gene Symbol
- MIR548XHG
- Entrez Gene ID
- 101927797
- Gene Name
- MIR548X host gene
- Gene Aliases
- Chromosome Location
- Chr. 21: 18561265 - 18759812 on Build GRCh38
- UniGene ID
- Hs.551743
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_109925.1 |
|
4 |
393 |
|
GenBank mRNA |
BC028044.1 |
- |
4 |
461 |
|
- Gene Symbol
- TDRKH
- Entrez Gene ID
- 11022
- Gene Name
- tudor and KH domain containing
- Gene Aliases
- TDRD2
- Chromosome Location
- Chr. 1: 151769354 - 151790576 on Build GRCh38
- UniGene ID
- Hs.144439
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK225160.1 |
- |
15 |
3337 |
|
View Full Product Details