Gene Transcripts
- Gene Symbol
- CEACAM1
- Entrez Gene ID
- 634
- Gene Name
- carcinoembryonic antigen related cell adhesion molecule 1
- Gene Aliases
- BGP, BGP1, BGPI
- Chromosome Location
- Chr. 19: 42507306 - 42528509 on Build GRCh38
- UniGene ID
- Hs.512682
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M76744.1 |
- |
2 |
104 |
|
- Gene Symbol
- OLAH
- Entrez Gene ID
- 55301
- Gene Name
- oleoyl-ACP hydrolase
- Gene Aliases
- AURA1, SAST, THEDC1
- Chromosome Location
- Chr. 10: 15043896 - 15073852 on Build GRCh38
- UniGene ID
- Hs.24309
- Species
- Homo sapiens
- Gene Symbol
- SARDH
- Entrez Gene ID
- 1757
- Gene Name
- sarcosine dehydrogenase
- Gene Aliases
- BPR-2, DMGDHL1, SAR, SARD, SDH
- Chromosome Location
- Chr. 9: 133663074 - 133739958 on Build GRCh38
- UniGene ID
- Hs.198003
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF095737.2 |
- |
7 |
1362 |
|
- Gene Symbol
- SIMC1
- Entrez Gene ID
- 375484
- Gene Name
- SUMO interacting motifs containing 1
- Gene Aliases
- C5orf25, OOMA1, PLEIAD
- Chromosome Location
- Chr. 5: 176238359 - 176345991 on Build GRCh38
- UniGene ID
- Hs.719847
- Species
- Homo sapiens
View Full Product Details