Gene Transcripts
- Gene Symbol
- RPS26
- Entrez Gene ID
- 6231
- Gene Name
- ribosomal protein S26
- Gene Aliases
- DBA10, S26
- Chromosome Location
- Chr. 12: 56041902 - 56044223 on Build GRCh38
- UniGene ID
- Hs.447562
- Species
- Homo sapiens
- Gene Symbol
- RPS26P11
- Entrez Gene ID
- 441502
- Gene Name
- ribosomal protein S26 pseudogene 11
- Gene Aliases
- RPS26L1, RPS26_22_1784, bA366E13.1
- Chromosome Location
- Chr. X: 72044409 - 72044974 on Build GRCh38
- UniGene ID
- Hs.449523
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002309.1 |
|
1 |
201 |
|
GenBank mRNA |
AW972305.1 |
- |
1 |
311 |
|
View Full Product Details