Gene Transcripts
- Gene Symbol
- JAM2
- Entrez Gene ID
- 58494
- Gene Name
- junctional adhesion molecule 2
- Gene Aliases
- C21orf43, CD322, JAM-B, JAMB, PRO245, VE-JAM, VEJAM
- Chromosome Location
- Chr. 21: 25639282 - 25717562 on Build GRCh38
- UniGene ID
- Hs.517227
- Species
- Homo sapiens
- Gene Symbol
- ABCC10
- Entrez Gene ID
- 89845
- Gene Name
- ATP binding cassette subfamily C member 10
- Gene Aliases
- EST182763, MRP7, SIMRP7
- Chromosome Location
- Chr. 6: 43427538 - 43451994 on Build GRCh38
- UniGene ID
- Hs.55879
- Species
- Homo sapiens
- Gene Symbol
- ACKR2
- Entrez Gene ID
- 1238
- Gene Name
- atypical chemokine receptor 2
- Gene Aliases
- CCBP2, CCR10, CCR9, CMKBR9, D6, hD6
- Chromosome Location
- Chr. 3: 42809472 - 42867283 on Build GRCh38
- UniGene ID
- Hs.146346
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC011588.1 |
- |
4 |
2988 |
|
BC018716.1 |
- |
4 |
2988 |
|
- Gene Symbol
- AGPAT3
- Entrez Gene ID
- 56894
- Gene Name
- 1-acylglycerol-3-phosphate O-acyltransferase 3
- Gene Aliases
- LPAAT-GAMMA1, LPAAT3
- Chromosome Location
- Chr. 21: 43865160 - 43987594 on Build GRCh38
- UniGene ID
- Hs.248785
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY358704.1 |
- |
|
2624 |
|
- Gene Symbol
- ASTN2
- Entrez Gene ID
- 23245
- Gene Name
- astrotactin 2
- Gene Aliases
- bA67K19.1
- Chromosome Location
- Chr. 9: 116425225 - 117415039 on Build GRCh38
- UniGene ID
- Hs.601562
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX648673.1 |
- |
1 |
4545 |
|
- Gene Symbol
- ATR
- Entrez Gene ID
- 545
- Gene Name
- ATR serine/threonine kinase
- Gene Aliases
- FCTCS, FRP1, MEC1, SCKL, SCKL1
- Chromosome Location
- Chr. 3: 142449235 - 142578826 on Build GRCh38
- UniGene ID
- Hs.271791
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB208847.1 |
- |
9 |
1789 |
|
- Gene Symbol
- CENPK
- Entrez Gene ID
- 64105
- Gene Name
- centromere protein K
- Gene Aliases
- AF5alpha, CENP-K, FKSG14, P33, Solt
- Chromosome Location
- Chr. 5: 65488919 - 65563194 on Build GRCh38
- UniGene ID
- Hs.529778
- Species
- Homo sapiens
- Gene Symbol
- CENPL
- Entrez Gene ID
- 91687
- Gene Name
- centromere protein L
- Gene Aliases
- C1orf155, CENP-L, dJ383J4.3
- Chromosome Location
- Chr. 1: 173799550 - 173824639 on Build GRCh38
- UniGene ID
- Hs.531856
- Species
- Homo sapiens
- Gene Symbol
- CEP57L1
- Entrez Gene ID
- 285753
- Gene Name
- centrosomal protein 57 like 1
- Gene Aliases
- C6orf182, bA487F23.2, cep57R
- Chromosome Location
- Chr. 6: 109095130 - 109167696 on Build GRCh38
- UniGene ID
- Hs.632616
- Species
- Homo sapiens
- Gene Symbol
- CHGB
- Entrez Gene ID
- 1114
- Gene Name
- chromogranin B
- Gene Aliases
- SCG1
- Chromosome Location
- Chr. 20: 5911328 - 5925361 on Build GRCh38
- UniGene ID
- Hs.516874
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK054871.1 |
- |
1 |
278 |
|
- Gene Symbol
- CPXM2
- Entrez Gene ID
- 119587
- Gene Name
- carboxypeptidase X (M14 family), member 2
- Gene Aliases
- CPX2, UNQ676
- Chromosome Location
- Chr. 10: 123745636 - 123944094 on Build GRCh38
- UniGene ID
- Hs.656887
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC027623.1 |
- |
10 |
1001 |
|
- Gene Symbol
- CRCP
- Entrez Gene ID
- 27297
- Gene Name
- CGRP receptor component
- Gene Aliases
- CGRP-RCP, CGRPRCP, RCP, RCP9
- Chromosome Location
- Chr. 7: 66114818 - 66154568 on Build GRCh38
- UniGene ID
- Hs.300684
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024548.1 |
|
3 |
222 |
|
GenBank mRNA |
BC052570.1 |
- |
3 |
436 |
|
DA955978.1 |
- |
3 |
160 |
|
- Gene Symbol
- DFNA5
- Entrez Gene ID
- 1687
- Gene Name
- DFNA5, deafness associated tumor suppressor
- Gene Aliases
- ICERE-1
- Chromosome Location
- Chr. 7: 24698355 - 24758020 on Build GRCh38
- UniGene ID
- Hs.520708
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094714.1 |
- |
2 |
441 |
|
- Gene Symbol
- FASTKD1
- Entrez Gene ID
- 79675
- Gene Name
- FAST kinase domains 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 169527879 - 169573921 on Build GRCh38
- UniGene ID
- Hs.529276
- Species
- Homo sapiens
- Gene Symbol
- FLJ41327
- Entrez Gene ID
- 401045
- Gene Name
- FLJ41327 protein
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.645461
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123321.1 |
- |
1 |
752 |
|
- Gene Symbol
- GRTP1
- Entrez Gene ID
- 79774
- Gene Name
- growth hormone regulated TBC protein 1
- Gene Aliases
- TBC1D6
- Chromosome Location
- Chr. 13: 113324164 - 113364158 on Build GRCh38
- UniGene ID
- Hs.745043
- Species
- Homo sapiens
- Gene Symbol
- KDSR
- Entrez Gene ID
- 2531
- Gene Name
- 3-ketodihydrosphingosine reductase
- Gene Aliases
- DHSR, FVT1, SDR35C1
- Chromosome Location
- Chr. 18: 63327736 - 63367273 on Build GRCh38
- UniGene ID
- Hs.74050
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126679.1 |
- |
1 |
624 |
|
- Gene Symbol
- KLRG1
- Entrez Gene ID
- 10219
- Gene Name
- killer cell lectin like receptor G1
- Gene Aliases
- 2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE
- Chromosome Location
- Chr. 12: 8989079 - 9255692 on Build GRCh38
- UniGene ID
- Hs.558446
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI433079.1 |
- |
1 |
41 |
|
- Gene Symbol
- LAMP3
- Entrez Gene ID
- 27074
- Gene Name
- lysosomal associated membrane protein 3
- Gene Aliases
- CD208, DC LAMP, DC-LAMP, DCLAMP, LAMP, LAMP-3, TSC403
- Chromosome Location
- Chr. 3: 183122213 - 183162879 on Build GRCh38
- UniGene ID
- Hs.518448
- Species
- Homo sapiens
- Gene Symbol
- LINC00999
- Entrez Gene ID
- 399744
- Gene Name
- long intergenic non-protein coding RNA 999
- Gene Aliases
- Chromosome Location
- Chr. 10: 38428146 - 38452153 on Build GRCh38
- UniGene ID
- Hs.497951
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024497.2 |
|
3 |
302 |
|
GenBank mRNA |
AK293765.1 |
- |
3 |
302 |
|
- Gene Symbol
- LINC01347
- Entrez Gene ID
- 731275
- Gene Name
- long intergenic non-protein coding RNA 1347
- Gene Aliases
- Chromosome Location
- Chr. 1: 243056314 - 243101744 on Build GRCh38
- UniGene ID
- Hs.722350
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_029401.1 |
|
11, 12 |
2053 |
|
GenBank mRNA |
AL137733.1 |
- |
11, 12 |
2053 |
|
- Gene Symbol
- LINGO2
- Entrez Gene ID
- 158038
- Gene Name
- leucine rich repeat and Ig domain containing 2
- Gene Aliases
- LERN3, LRRN6C
- Chromosome Location
- Chr. 9: 27937617 - 29213981 on Build GRCh38
- UniGene ID
- Hs.444665
- Species
- Homo sapiens
- Gene Symbol
- LOC100130419
- Entrez Gene ID
- 100130419
- Gene Name
- uncharacterized LOC100130419
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.651773
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126781.1 |
- |
2 |
2307 |
|
- Gene Symbol
- LOC100134822
- Entrez Gene ID
- 100134822
- Gene Name
- uncharacterized LOC100134822
- Gene Aliases
- Chromosome Location
- Chr. 1: 497259 - 532779 on Build GRCh38
- UniGene ID
- Hs.744675
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL137655.1 |
- |
10 |
1468 |
|
- Gene Symbol
- LOC100506974
- Entrez Gene ID
- 100506974
- Gene Name
- uncharacterized LOC100506974
- Gene Aliases
- Chromosome Location
- Chr. 17: 13790327 - 13898625 on Build GRCh38
- UniGene ID
- Hs.643090
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123263.1 |
- |
1 |
386 |
|
- Gene Symbol
- LOC101928295
- Entrez Gene ID
- 101928295
- Gene Name
- uncharacterized LOC101928295
- Gene Aliases
- Chromosome Location
- Chr. 19: 49368705 - 49388081 on Build GRCh38
- UniGene ID
- Hs.640180
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110729.1 |
|
2 |
754 |
|
GenBank mRNA |
AK126060.1 |
- |
2 |
752 |
|
- Gene Symbol
- LOC105371998
- Entrez Gene ID
- 105371998
- Gene Name
- uncharacterized LOC105371998
- Gene Aliases
- Chromosome Location
- Chr. 18: 12432774 - 12447798 on Build GRCh38
- UniGene ID
- Hs.515283
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK128548.1 |
- |
1 |
2764 |
|
- Gene Symbol
- LOC105376896
- Entrez Gene ID
- 105376896
- Gene Name
- uncharacterized LOC105376896
- Gene Aliases
- Chromosome Location
- Chr. 1: 28239511 - 28247324 on Build GRCh38
- UniGene ID
- Hs.523897
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095431.1 |
- |
1 |
1462 |
|
- Gene Symbol
- LOC105379811
- Entrez Gene ID
- 105379811
- Gene Name
- uncharacterized LOC105379811
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC008001.1 |
- |
1 |
1021 |
|
- Gene Symbol
- LOC107984814
- Entrez Gene ID
- 107984814
- Gene Name
- uncharacterized LOC107984814
- Gene Aliases
- Chromosome Location
- Chr. 16: 72664780 - 72752034 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- MED15
- Entrez Gene ID
- 51586
- Gene Name
- mediator complex subunit 15
- Gene Aliases
- ARC105, CAG7A, CTG7A, PCQAP, TIG-1, TIG1, TNRC7
- Chromosome Location
- Chr. 22: 20507542 - 20587632 on Build GRCh38
- UniGene ID
- Hs.517421
- Species
- Homo sapiens
- Gene Symbol
- NEDD8
- Entrez Gene ID
- 4738
- Gene Name
- neural precursor cell expressed, developmentally down-regulated 8
- Gene Aliases
- NEDD-8
- Chromosome Location
- Chr. 14: 24216851 - 24232370 on Build GRCh38
- UniGene ID
- Hs.531064
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125214.1 |
- |
2 |
451 |
|
- Gene Symbol
- NPC1L1
- Entrez Gene ID
- 29881
- Gene Name
- NPC1 like intracellular cholesterol transporter 1
- Gene Aliases
- NPC11L1
- Chromosome Location
- Chr. 7: 44512440 - 44541650 on Build GRCh38
- UniGene ID
- Hs.567486
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013389.2 |
NP_037521.2
|
15 |
3225 |
|
GenBank mRNA |
AF192522.1 |
- |
15 |
3225 |
|
- Gene Symbol
- PDCL
- Entrez Gene ID
- 5082
- Gene Name
- phosducin like
- Gene Aliases
- PhLP
- Chromosome Location
- Chr. 9: 122816058 - 122828656 on Build GRCh38
- UniGene ID
- Hs.271749
- Species
- Homo sapiens
- Gene Symbol
- PLEKHG2
- Entrez Gene ID
- 64857
- Gene Name
- pleckstrin homology and RhoGEF domain containing G2
- Gene Aliases
- ARHGEF42, CLG, LDAMD
- Chromosome Location
- Chr. 19: 39412582 - 39428415 on Build GRCh38
- UniGene ID
- Hs.631574
- Species
- Homo sapiens
- Gene Symbol
- PPP1R12A
- Entrez Gene ID
- 4659
- Gene Name
- protein phosphatase 1 regulatory subunit 12A
- Gene Aliases
- M130, MBS, MYPT1
- Chromosome Location
- Chr. 12: 79773563 - 79935455 on Build GRCh38
- UniGene ID
- Hs.49582
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125671.1 |
- |
1 |
577 |
|
- Gene Symbol
- PTPN21
- Entrez Gene ID
- 11099
- Gene Name
- protein tyrosine phosphatase, non-receptor type 21
- Gene Aliases
- PTPD1, PTPRL10
- Chromosome Location
- Chr. 14: 88465778 - 88554779 on Build GRCh38
- UniGene ID
- Hs.437040
- Species
- Homo sapiens
- Gene Symbol
- RBM14
- Entrez Gene ID
- 10432
- Gene Name
- RNA binding motif protein 14
- Gene Aliases
- COAA, PSP2, SIP, SYTIP1, TMEM137
- Chromosome Location
- Chr. 11: 66616582 - 66629934 on Build GRCh38
- UniGene ID
- Hs.533712
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209007.1 |
- |
1 |
394 |
|
- Gene Symbol
- RNF125
- Entrez Gene ID
- 54941
- Gene Name
- ring finger protein 125
- Gene Aliases
- TNORS, TRAC-1, TRAC1
- Chromosome Location
- Chr. 18: 32018203 - 32090751 on Build GRCh38
- UniGene ID
- Hs.633703
- Species
- Homo sapiens
- Gene Symbol
- SLC1A2
- Entrez Gene ID
- 6506
- Gene Name
- solute carrier family 1 member 2
- Gene Aliases
- EAAT2, GLT-1, HBGT
- Chromosome Location
- Chr. 11: 35251205 - 35420063 on Build GRCh38
- UniGene ID
- Hs.502338
- Species
- Homo sapiens
- Gene Symbol
- STYX
- Entrez Gene ID
- 6815
- Gene Name
- serine/threonine/tyrosine interacting protein
- Gene Aliases
- Chromosome Location
- Chr. 14: 52730159 - 52774989 on Build GRCh38
- UniGene ID
- Hs.364980
- Species
- Homo sapiens
- Gene Symbol
- SYT14
- Entrez Gene ID
- 255928
- Gene Name
- synaptotagmin 14
- Gene Aliases
- SCAR11, sytXIV
- Chromosome Location
- Chr. 1: 209938165 - 210171315 on Build GRCh38
- UniGene ID
- Hs.658866
- Species
- Homo sapiens
- Gene Symbol
- TBL1X
- Entrez Gene ID
- 6907
- Gene Name
- transducin (beta)-like 1X-linked
- Gene Aliases
- EBI, SMAP55, TBL1
- Chromosome Location
- Chr. X: 9463295 - 9719740 on Build GRCh38
- UniGene ID
- Hs.495656
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC029394.1 |
- |
3 |
675 |
|
- Gene Symbol
- TRIM65
- Entrez Gene ID
- 201292
- Gene Name
- tripartite motif containing 65
- Gene Aliases
- 4732463G12Rik
- Chromosome Location
- Chr. 17: 75880379 - 75897008 on Build GRCh38
- UniGene ID
- Hs.189823
- Species
- Homo sapiens
- Gene Symbol
- VRK2
- Entrez Gene ID
- 7444
- Gene Name
- vaccinia related kinase 2
- Gene Aliases
- Chromosome Location
- Chr. 2: 57907651 - 58163991 on Build GRCh38
- UniGene ID
- Hs.715298
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK307113.1 |
- |
2 |
216 |
|
- Gene Symbol
- YBX1
- Entrez Gene ID
- 4904
- Gene Name
- Y-box binding protein 1
- Gene Aliases
- BP-8, CBF-A, CSDA2, CSDB, DBPB, EFI-A, MDR-NF1, NSEP-1, NSEP1, YB-1, YB1
- Chromosome Location
- Chr. 1: 42682235 - 42703803 on Build GRCh38
- UniGene ID
- Hs.473583
- Species
- Homo sapiens
- Gene Symbol
- ZNF34
- Entrez Gene ID
- 80778
- Gene Name
- zinc finger protein 34
- Gene Aliases
- KOX32
- Chromosome Location
- Chr. 8: 144772224 - 144787371 on Build GRCh38
- UniGene ID
- Hs.631854
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
W43004.1 |
- |
1 |
380 |
|
- Gene Symbol
- ZNF492
- Entrez Gene ID
- 57615
- Gene Name
- zinc finger protein 492
- Gene Aliases
- ZNF115
- Chromosome Location
- Chr. 19: 22634324 - 22667670 on Build GRCh38
- UniGene ID
- Hs.232108
- Species
- Homo sapiens
- Gene Symbol
- ZNF571
- Entrez Gene ID
- 51276
- Gene Name
- zinc finger protein 571
- Gene Aliases
- HSPC059
- Chromosome Location
- Chr. 19: 37564253 - 37594830 on Build GRCh38
- UniGene ID
- Hs.590944
- Species
- Homo sapiens
View Full Product Details