Gene Transcripts
- Gene Symbol
- PDPK1
- Entrez Gene ID
- 5170
- Gene Name
- 3-phosphoinositide dependent protein kinase 1
- Gene Aliases
- PDK1, PDPK2, PDPK2P, PRO0461
- Chromosome Location
- Chr. 16: 2537964 - 2603190 on Build GRCh38
- UniGene ID
- Hs.459691
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF111845.2 |
- |
1 |
903 |
|
View Full Product Details