Gene Transcripts
- Gene Symbol
- JAM2
- Entrez Gene ID
- 58494
- Gene Name
- junctional adhesion molecule 2
- Gene Aliases
- C21orf43, CD322, JAM-B, JAMB, PRO245, VE-JAM, VEJAM
- Chromosome Location
- Chr. 21: 25639282 - 25717562 on Build GRCh38
- UniGene ID
- Hs.517227
- Species
- Homo sapiens
- Gene Symbol
- FASTKD1
- Entrez Gene ID
- 79675
- Gene Name
- FAST kinase domains 1
- Gene Aliases
- Chromosome Location
- Chr. 2: 169527879 - 169573921 on Build GRCh38
- UniGene ID
- Hs.529276
- Species
- Homo sapiens
- Gene Symbol
- LOC100507494
- Entrez Gene ID
- 100507494
- Gene Name
- uncharacterized LOC100507494
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.683963
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK090481.1 |
- |
|
1614 |
|
- Gene Symbol
- LOC105379811
- Entrez Gene ID
- 105379811
- Gene Name
- uncharacterized LOC105379811
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC008001.1 |
- |
1 |
1028 |
|
- Gene Symbol
- STYX
- Entrez Gene ID
- 6815
- Gene Name
- serine/threonine/tyrosine interacting protein
- Gene Aliases
- Chromosome Location
- Chr. 14: 52730159 - 52774989 on Build GRCh38
- UniGene ID
- Hs.364980
- Species
- Homo sapiens
- Gene Symbol
- TRPM7
- Entrez Gene ID
- 54822
- Gene Name
- transient receptor potential cation channel subfamily M member 7
- Gene Aliases
- ALSPDC, CHAK, CHAK1, LTRPC7, LTrpC-7, TRP-PLIK
- Chromosome Location
- Chr. 15: 50557158 - 50686835 on Build GRCh38
- UniGene ID
- Hs.512894
- Species
- Homo sapiens
- Gene Symbol
- YBX1
- Entrez Gene ID
- 4904
- Gene Name
- Y-box binding protein 1
- Gene Aliases
- BP-8, CBF-A, CSDA2, CSDB, DBPB, EFI-A, MDR-NF1, NSEP-1, NSEP1, YB-1, YB1
- Chromosome Location
- Chr. 1: 42682235 - 42703803 on Build GRCh38
- UniGene ID
- Hs.473583
- Species
- Homo sapiens
View Full Product Details