Gene Transcripts
- Gene Symbol
- RPS27A
- Entrez Gene ID
- 6233
- Gene Name
- ribosomal protein S27a
- Gene Aliases
- CEP80, HEL112, S27A, UBA80, UBC, UBCEP1, UBCEP80
- Chromosome Location
- Chr. 2: 55231903 - 55235853 on Build GRCh38
- UniGene ID
- Hs.311640
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135592.2 |
NP_001129064.1
|
3, 4 |
320 |
|
NM_001177413.1 |
NP_001170884.1
|
2, 3 |
244 |
|
NM_002954.5 |
NP_002945.1
|
3, 4 |
418 |
|
GenBank mRNA |
AK225079.1 |
- |
1 |
135 |
|
AK225718.1 |
- |
1 |
135 |
|
AK315695.1 |
- |
3, 4 |
135 |
|
AY952889.1 |
- |
1 |
122 |
|
BC001392.2 |
- |
1 |
124 |
3050927 |
BC042362.1 |
- |
3, 4 |
135 |
4831143 |
BC053371.1 |
- |
1 |
124 |
5531043 |
BC066293.1 |
- |
1 |
135 |
30336950 |
BF678026.1 |
- |
2, 3 |
243 |
|
BQ276758.1 |
- |
2, 3 |
138 |
|
CK004647.1 |
- |
3, 4 |
294 |
|
CN344335.1 |
- |
2, 3 |
119 |
|
CR746519.1 |
- |
3, 4 |
429 |
|
DA527860.1 |
- |
2, 3 |
220 |
|
DB505702.1 |
- |
3, 4 |
320 |
|
EU794671.1 |
- |
3, 4 |
135 |
|
M10939.1 |
- |
2, 3 |
85 |
|
S79522.1 |
- |
3, 4 |
135 |
|
X63237.1 |
- |
1 |
122 |
|
View Full Product Details