Gene Transcripts
- Gene Symbol
- EYS
- Entrez Gene ID
- 346007
- Gene Name
- eyes shut homolog (Drosophila)
- Gene Aliases
- C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
- Chromosome Location
- Chr. 6: 63719980 - 65707225 on Build GRCh38
- UniGene ID
- Hs.25067
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX647741.1 |
- |
5 |
4877 |
|
- Gene Symbol
- LINC00504
- Entrez Gene ID
- 201853
- Gene Name
- long intergenic non-protein coding RNA 504
- Gene Aliases
- Chromosome Location
- Chr. 4: 14470465 - 14888169 on Build GRCh38
- UniGene ID
- Hs.551051
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_126435.1 |
|
7 |
2016 |
|
GenBank mRNA |
BC036758.2 |
- |
7 |
2028 |
|
View Full Product Details