Gene Transcripts

Gene Symbol
ESRRG
Entrez Gene ID
2104
Gene Name
estrogen related receptor gamma
Gene Aliases
ERR3, ERRgamma, NR3B3
Chromosome Location
Chr. 1: 216503246 - 217137757 on Build GRCh38
UniGene ID
Hs.444225
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134285.2 NP_001127757.1 5 757
NM_001243506.1 NP_001230435.1 3 267
NM_001243509.1 NP_001230438.1 5 1135
NM_001243510.2 NP_001230439.1 5 751
NM_001243511.2 NP_001230440.1 4 633
NM_001243512.1 NP_001230441.1 5 700
NM_001243513.1 NP_001230442.1 4 720
NM_001243514.1 NP_001230443.1 3 877
NM_001243515.1 NP_001230444.1 5 976
NM_001243518.1 NP_001230447.1 4 809
NM_001243519.1 NP_001230448.1 5 980
NM_001438.3 NP_001429.2 3 681
NM_206594.2 NP_996317.1 5 801
NM_206595.2 NP_996318.1 4 683
XM_011509265.2 XP_011507567.1 5 4744
XM_011509266.2 XP_011507568.1 5 1138
XM_011509267.1 XP_011507569.1 6 1061
XM_011509268.2 XP_011507570.1 5 946
XM_011509269.2 XP_011507571.1 3 545
XM_011509270.1 XP_011507572.1 3 833
XM_011509271.2 XP_011507573.1 4 826
XM_011509274.1 XP_011507576.1 5 854
XM_011509275.1 XP_011507577.1 5 810
XM_011509276.1 XP_011507578.1 5 969
XM_011509277.1 XP_011507579.1 4 661
XM_011509278.1 XP_011507580.1 4 692
XM_011509279.1 XP_011507581.1 4 837
XM_011509280.2 XP_011507582.1 4 814
XM_017000621.1 XP_016856110.1 6 4902
XM_017000622.1 XP_016856111.1 5 1134
XM_017000623.1 XP_016856112.1 5 1094
XM_017000624.1 XP_016856113.1 5 3460
XM_017000625.1 XP_016856114.1 5 997
XM_017000626.1 XP_016856115.1 5 875
XM_017000627.1 XP_016856116.1 5 938
XM_017000628.1 XP_016856117.1 7 1146
XM_017000629.1 XP_016856118.1 4 690
XM_017000630.1 XP_016856119.1 7 1192
XM_017000631.1 XP_016856120.1 6 939
XM_017000632.1 XP_016856121.1 7 1118
XM_017000633.1 XP_016856122.1 5 779
XM_017000634.1 XP_016856123.1 5 821
XM_017000635.1 XP_016856124.1 7 3896
XM_017000636.1 XP_016856125.1 6 965
XM_017000637.1 XP_016856126.1 6 1066
XM_017000638.1 XP_016856127.1 6 939
XM_017000639.1 XP_016856128.1 5 779
XM_017000640.1 XP_016856129.1 4 3042
XM_017000641.1 XP_016856130.1 4 837
XM_017000642.1 XP_016856131.1 5 935
XM_017000643.1 XP_016856132.1 6 932
XM_017000644.1 XP_016856133.1 5 810
XM_017000645.1 XP_016856134.1 4 692
XM_017000646.1 XP_016856135.1 5 3175
XM_017000647.1 XP_016856136.1 4 924
XM_017000648.1 XP_016856137.1 4 813
XM_017000649.1 XP_016856138.1 6 1261
GenBank mRNA AB020639.1 - 3 662
AB307715.1 - 2 439
AB362218.1 - 5 1054
AF058291.1 - 4 657
AF094518.1 - 3 633
AK290945.1 - 5 957
AK291028.1 - 4 658
AK291647.1 - 3 654
AK298851.1 - 3 234
AY388456.1 - 3 636
AY388457.1 - 4 669
AY388458.1 - 5 695
AY388459.1 - 4 712
AY388460.1 - 3 552
AY388461.1 - 3 926
BC008218.2 - 5 840
BC064700.1 - 4 790 30344716
BX647076.1 - 5 757
CR749497.1 - 5 750
HQ692853.1 - 3 508
HQ692854.1 - 2 439

View Full Product Details