Gene Transcripts

Gene Symbol
ATP7A
Entrez Gene ID
538
Gene Name
ATPase copper transporting alpha
Gene Aliases
DSMAX, MK, MNK, SMAX3
Chromosome Location
Chr. X: 77910656 - 78050395 on Build GRCh38
UniGene ID
Hs.496414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000052.6 NP_000043.4 10 2469
GenBank mRNA AB117973.1 - 10 2396
L06133.1 - 10 2413

View Full Product Details