Gene Transcripts

Gene Symbol
ATP7A
Entrez Gene ID
538
Gene Name
ATPase copper transporting alpha
Gene Aliases
DSMAX, MK, MNK, SMAX3
Chromosome Location
Chr. X: 77910656 - 78050395 on Build GRCh38
UniGene ID
Hs.496414
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000052.6 NP_000043.4 11 2628
NM_001282224.1 NP_001269153.1 10 2394
GenBank mRNA AB117973.1 - 11 2555
L06133.1 - 11 2572

View Full Product Details