Gene Transcripts

Gene Symbol
AAK1
Entrez Gene ID
22848
Gene Name
AP2 associated kinase 1
Gene Aliases
Chromosome Location
Chr. 2: 69457995 - 69643845 on Build GRCh38
UniGene ID
Hs.468878
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014911.3 NP_055726.3 22 13983
GenBank mRNA AK127187.1 - 1 842
BC132937.1 - 1 828
BC132939.1 - 1 828

View Full Product Details