Gene Transcripts

Gene Symbol
BTBD2
Entrez Gene ID
55643
Gene Name
BTB domain containing 2
Gene Aliases
Chromosome Location
Chr. 19: 1985438 - 2015703 on Build GRCh38
UniGene ID
Hs.465543
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017797.3 NP_060267.2 9 1568
XM_005259593.1 XP_005259650.1 9 1333
XM_011528127.1 XP_011526429.1 7 934
GenBank mRNA AF355797.1 - 9 1568
AK000393.1 - 6 1362
AK027481.1 - 7 930
AK056818.1 - 3 1703
AK225831.1 - 9 1567
AL360275.1 - 6 799
AY007116.1 - 9 1290
BC000564.2 - 6 858
BC008035.2 - 6 852
BC053986.1 - 5 753
BC069199.1 - 7 994
BC094820.1 - 9 1302

View Full Product Details