Gene Transcripts

Gene Symbol
Wee1
Entrez Gene ID
308937
Gene Name
WEE1 G2 checkpoint kinase
Gene Aliases
Chromosome Location
Chr. 1: 174765230 - 174790863 on Build Rnor_6.0
UniGene ID
Rn.208255
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012742.1 NP_001012760.1 11 2208
GenBank mRNA BC090346.1 - 11 2208
D31838.1 - 11 2407

View Full Product Details