Gene Transcripts
- Gene Symbol
- ACER3
- Entrez Gene ID
- 55331
- Gene Name
- alkaline ceramidase 3
- Gene Aliases
- APHC, PHCA
- Chromosome Location
- Chr. 11: 76860873 - 77026797 on Build GRCh38
- UniGene ID
- Hs.23862
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300953.1 |
NP_001287882.1
|
5, 6 |
439 |
|
NM_001300954.1 |
NP_001287883.1
|
6, 7 |
621 |
|
NM_001300955.1 |
NP_001287884.1
|
5, 6 |
497 |
|
NM_018367.6 |
NP_060837.3
|
6, 7 |
550 |
|
XM_011545151.2 |
XP_011543453.1
|
6, 7 |
516 |
|
XM_011545152.2 |
XP_011543454.1
|
6, 7 |
486 |
|
XM_011545153.2 |
XP_011543455.1
|
6, 7 |
516 |
|
XM_017017988.1 |
XP_016873477.1
|
6, 7 |
516 |
|
GenBank mRNA |
AF214454.1 |
- |
6, 7 |
504 |
|
AF327353.1 |
- |
6, 7 |
515 |
|
AK002100.1 |
- |
8, 9 |
681 |
|
AK293800.1 |
- |
7, 8 |
521 |
|
AK294978.1 |
- |
5, 6 |
370 |
|
AK295142.1 |
- |
6, 7 |
538 |
|
AK295327.1 |
- |
7, 8 |
634 |
|
AK308856.1 |
- |
8, 9 |
678 |
|
AK315000.1 |
- |
6, 7 |
516 |
|
AK316120.1 |
- |
5, 6 |
435 |
|
BC063034.1 |
- |
7, 8 |
510 |
|
BC073853.1 |
- |
6, 7 |
467 |
3867010 |
BG702017.1 |
- |
6, 7 |
555 |
|
View Full Product Details