Gene Transcripts

Gene Symbol
TENM2
Entrez Gene ID
57451
Gene Name
teneurin transmembrane protein 2
Gene Aliases
ODZ2, TEN-M2, TNM2, ten-2
Chromosome Location
Chr. 5: 166979066 - 168264159 on Build GRCh38
UniGene ID
Hs.631957
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122679.1 NP_001116151.1 1 79
XM_005265950.2 XP_005266007.1 3 726
XM_006714897.2 XP_006714960.1 3 726
XM_011534604.2 XP_011532906.1 3 726
XM_017009660.1 XP_016865149.1 3 726
XM_017009661.1 XP_016865150.1 3 726
XM_017009662.1 XP_016865151.1 3 726
GenBank mRNA HE578282.1 - 1 79
HE578283.1 - 1 79

View Full Product Details