Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001370.1 | NP_001361.1 | 19 | 3068 | |
XM_006711956.2 | XP_006712019.1 | 19 | 3173 | ||
XM_011532649.2 | XP_011530951.1 | 19 | 3022 | ||
XM_011532650.2 | XP_011530952.1 | 19 | 3023 | ||
XM_011532652.1 | XP_011530954.1 | 19 | 3068 | ||
XM_011532653.1 | XP_011530955.1 | 19 | 3068 | ||
XM_011532654.1 | XP_011530956.1 | 18 | 2801 | ||
XM_011532655.2 | XP_011530957.1 | 15 | 2090 | ||
XM_011532657.1 | XP_011530959.1 | 13 | 1829 | ||
XM_011532660.1 | XP_011530962.1 | 19 | 3068 | ||
XM_017003521.1 | XP_016859010.1 | 19 | 3068 | ||
XM_017003522.1 | XP_016859011.1 | 14 | 2894 | ||
XM_017003523.1 | XP_016859012.1 | 14 | 2061 | ||
XM_017003524.1 | XP_016859013.1 | 19 | 3068 |