Gene Transcripts

Gene Symbol
Olr1491
Entrez Gene ID
404969
Gene Name
olfactory receptor 1491
Gene Aliases
Chromosome Location
Chr. 10: 60405271 - 60406209 on Build Rnor_6.0
UniGene ID
Rn.142088
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000717.1 NP_001000717.1 1 23

View Full Product Details