Gene Transcripts
- Gene Symbol
- CRYZL1
- Entrez Gene ID
- 9946
- Gene Name
- crystallin zeta like 1
- Gene Aliases
- 4P11, QOH-1
- Chromosome Location
- Chr. 21: 33589341 - 33641854 on Build GRCh38
- UniGene ID
- Hs.352671
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145858.2 |
NP_665857.2
|
3, 4 |
332 |
|
GenBank mRNA |
AF029689.1 |
- |
3, 4 |
199 |
|
AF098798.1 |
- |
3, 4 |
199 |
|
AK001293.1 |
- |
3, 4 |
225 |
|
AK057604.1 |
- |
3, 4 |
816 |
|
AK315707.1 |
- |
3, 4 |
192 |
|
BC013155.1 |
- |
3, 4 |
200 |
|
BC033023.2 |
- |
3, 4 |
186 |
5269248 |
BI599245.1 |
- |
3, 4 |
237 |
|
BX647276.1 |
- |
3, 4 |
219 |
|
DB037192.1 |
- |
3, 4 |
333 |
|
View Full Product Details