Gene Transcripts
- Gene Symbol
- LYST
- Entrez Gene ID
- 1130
- Gene Name
- lysosomal trafficking regulator
- Gene Aliases
- CHS, CHS1
- Chromosome Location
- Chr. 1: 235661031 - 235883708 on Build GRCh38
- UniGene ID
- Hs.532411
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000081.3 |
NP_000072.2
|
4, 5 |
466 |
|
NM_001301365.1 |
NP_001288294.1
|
4, 5 |
903 |
|
XM_011544031.1 |
XP_011542333.1
|
4, 5 |
466 |
|
XM_011544032.1 |
XP_011542334.1
|
4, 5 |
869 |
|
XM_011544033.2 |
XP_011542335.1
|
4, 5 |
616 |
|
XM_011544034.1 |
XP_011542336.1
|
4, 5 |
466 |
|
XM_011544035.2 |
XP_011542337.1
|
4, 5 |
466 |
|
XM_011544037.2 |
XP_011542339.1
|
4, 5 |
466 |
|
XM_011544039.2 |
XP_011542341.1
|
4, 5 |
466 |
|
XM_011544040.2 |
XP_011542342.1
|
4, 5 |
466 |
|
XM_017000150.1 |
XP_016855639.1
|
4, 5 |
466 |
|
XM_017000151.1 |
XP_016855640.1
|
4, 5 |
466 |
|
GenBank mRNA |
AK307457.1 |
- |
4, 5 |
869 |
|
BC144702.1 |
- |
4, 5 |
767 |
|
U67615.1 |
- |
4, 5 |
473 |
|
U70064.1 |
- |
4, 5 |
836 |
|
U72192.1 |
- |
4, 5 |
836 |
|
U84744.1 |
- |
4, 5 |
836 |
|
View Full Product Details