Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005222.3 | NP_005213.3 | 2 | 484 | |
GenBank mRNA | AK094086.1 | - | 2 | 295 | |
BC069363.1 | - | 2 | 130 | 7262091 | |
BC103688.1 | - | 2 | 163 | 40000523 | |
BC103689.1 | - | 2 | 163 | 40000521 | |
BC103690.1 | - | 2 | 163 | 40000520 | |
BC109381.1 | - | 2 | 163 | 40000518 |