Gene Transcripts
- Gene Symbol
- CEBPA-AS1
- Entrez Gene ID
- 80054
- Gene Name
- CEBPA antisense RNA 1 (head to head)
- Gene Aliases
- Chromosome Location
- Chr. 19: 33302857 - 33305057 on Build GRCh38
- UniGene ID
- Hs.287521
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026887.1 |
|
1 |
2173 |
|
GenBank mRNA |
AK022417.1 |
- |
1 |
2075 |
|
BC027959.1 |
- |
1 |
2156 |
|
View Full Product Details