Gene Transcripts

Gene Symbol
C1QTNF4
Entrez Gene ID
114900
Gene Name
C1q and tumor necrosis factor related protein 4
Gene Aliases
CTRP4, ZACRP4
Chromosome Location
Chr. 11: 47589664 - 47596336 on Build GRCh38
UniGene ID
Hs.662633
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031909.2 NP_114115.2 2 1034
XM_017017165.1 XP_016872654.1 3 2352
XM_017017166.1 XP_016872655.1 3 2128
GenBank mRNA AF329838.1 - 2 991
BC035628.1 - 2 817 5199881

View Full Product Details