Gene Transcripts
- Gene Symbol
- SMCR5
- Entrez Gene ID
- 140771
- Gene Name
- Smith-Magenis syndrome chromosome region, candidate 5 (non-protein coding)
- Gene Aliases
- NCRNA00034
- Chromosome Location
- Chr. 17: 17776686 - 17779529 on Build GRCh38
- UniGene ID
- Hs.352643
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024007.1 |
|
1 |
833 |
|
GenBank mRNA |
AF467442.1 |
- |
1 |
833 |
|
View Full Product Details