Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_130760.2 | NP_570116.2 | 5 | 1261 | |
NM_130762.2 | NP_570118.1 | 4 | 1000 | ||
GenBank mRNA | AK092211.1 | - | 3 | 1496 | |
BC137506.1 | - | 4 | 1036 | 9021506 | |
BC137507.1 | - | 4 | 1036 | 9021507 | |
U82483.1 | - | 5 | 1261 |