Gene Transcripts

Gene Symbol
PITX3
Entrez Gene ID
5309
Gene Name
paired like homeodomain 3
Gene Aliases
ASMD, ASOD, CTPP4, CTRCT11, PTX3
Chromosome Location
Chr. 10: 102230189 - 102241474 on Build GRCh38
UniGene ID
Hs.137568
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005029.3 NP_005020.1 3 322
GenBank mRNA AF041339.1 - 2 171
BC011642.2 - 3 321 4131442

View Full Product Details