Gene Transcripts
- Gene Symbol
- Fas
- Entrez Gene ID
- 246097
- Gene Name
- Fas cell surface death receptor
- Gene Aliases
- Tnfrsf6
- Chromosome Location
- Chr. 1: 252589785 - 252624790 on Build Rnor_6.0
- UniGene ID
- Rn.162521
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
D26113.1 |
- |
2 |
130 |
|
View Full Product Details