Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012949.2 | NP_037081.2 | 8 | 776 | |
XM_006246599.2 | XP_006246661.1 | 8 | 847 | ||
XM_006246600.2 | XP_006246662.1 | 8 | 827 | ||
GenBank mRNA | BC083566.1 | - | 8 | 763 | |
FQ214810.1 | - | 7 | 754 | ||
FQ215034.1 | - | 4 | 453 | ||
FQ215310.1 | - | 4 | 454 | ||
FQ215357.1 | - | 8 | 798 | ||
FQ215526.1 | - | 8 | 786 | ||
FQ215607.1 | - | 4 | 453 | ||
FQ215648.1 | - | 8 | 786 | ||
FQ215759.1 | - | 8 | 798 | ||
FQ215780.1 | - | 4 | 453 | ||
FQ215843.1 | - | 7 | 754 | ||
FQ215865.1 | - | 8 | 786 | ||
FQ216150.1 | - | 7 | 755 | ||
FQ216271.1 | - | 4 | 454 | ||
FQ216382.1 | - | 8 | 788 | ||
FQ216660.1 | - | 7 | 754 | ||
FQ217095.1 | - | 8 | 786 | ||
FQ223831.1 | - | 4 | 453 | ||
FQ224938.1 | - | 8 | 786 | ||
Y00979.1 | - | 7 | 738 |