Gene Transcripts

Gene Symbol
Capza3
Entrez Gene ID
29324
Gene Name
capping protein (actin filament) muscle Z-line, alpha 3
Gene Aliases
Cappa3
Chromosome Location
Chr. 4: 174181644 - 174182857 on Build Rnor_6.0
UniGene ID
Rn.42865
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017164.2 NP_058860.2 1 265
GenBank mRNA BC078870.1 - 1 265
Y12538.1 - 1 245

View Full Product Details