Gene Transcripts

Gene Symbol
Rad50
Entrez Gene ID
64012
Gene Name
RAD50 homolog (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 10: 39002130 - 39054042 on Build Rnor_6.0
UniGene ID
Rn.51136
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022246.1 NP_071582.1 4 668
GenBank mRNA AF218576.1 - 4 668

View Full Product Details