Gene Transcripts

Gene Symbol
Nkx6-1
Entrez Gene ID
65193
Gene Name
NK6 homeobox 1
Gene Aliases
Nkx6.1, Nkx61, Nkx6a
Chromosome Location
Chr. 14: 9555264 - 9562197 on Build Rnor_6.0
UniGene ID
Rn.11387
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031737.1 NP_113925.1 3 1762
GenBank mRNA AF004431.1 - 3 1762

View Full Product Details