Gene Transcripts

Gene Symbol
Akt1
Entrez Gene ID
24185
Gene Name
v-akt murine thymoma viral oncogene homolog 1
Gene Aliases
Akt
Chromosome Location
Chr. 6: 137218398 - 137239970 on Build Rnor_6.0
UniGene ID
Rn.11422
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033230.2 NP_150233.1 4 413
XM_006240631.2 XP_006240693.1 5 912
XM_008764918.1 XP_008763140.1 4 2459
GenBank mRNA D30040.1 - 4 417

View Full Product Details