Gene Transcripts
- Gene Symbol
- Pi4k2a
- Entrez Gene ID
- 114554
- Gene Name
- phosphatidylinositol 4-kinase type 2 alpha
- Gene Aliases
- Pi4KII
- Chromosome Location
- Chr. 1: 261337594 - 261362509 on Build Rnor_6.0
- UniGene ID
- Rn.162533
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053735.1 |
NP_446187.1
|
1, 2 |
499 |
|
GenBank mRNA |
AY029199.1 |
- |
1, 2 |
499 |
|
View Full Product Details