Gene Transcripts

Gene Symbol
Ube2n
Entrez Gene ID
116725
Gene Name
ubiquitin-conjugating enzyme E2N
Gene Aliases
Chromosome Location
Chr. 7: 36610147 - 36640190 on Build Rnor_6.0
UniGene ID
Rn.230134
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053928.2 NP_446380.1 2 491
GenBank mRNA AB032739.1 - 2 152
BC090072.1 - 2 491 7113593

View Full Product Details