Gene Transcripts

Gene Symbol
Slc17a7
Entrez Gene ID
116638
Gene Name
solute carrier family 17 (vesicular glutamate transporter), member 7
Gene Aliases
BNPI, Vglut1
Chromosome Location
Chr. 1: 101161265 - 101172292 on Build Rnor_6.0
UniGene ID
Rn.10267
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053859.2 NP_446311.1 6 825
GenBank mRNA EU253553.1 - 7 790
FQ211811.1 - 838
U07609.1 - 6 828

View Full Product Details