Gene Transcripts

Gene Symbol
TFDP3
Entrez Gene ID
51270
Gene Name
transcription factor Dp family member 3
Gene Aliases
CT30, DP4, HCA661
Chromosome Location
Chr. X: 133216669 - 133218348 on Build GRCh38
UniGene ID
Hs.142908
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016521.2 NP_057605.3 1 1099
GenBank mRNA AF219119.2 - 1 1099
BC074762.2 - 1 858
BC117336.1 - 1 895
BC126361.1 - 1 895

View Full Product Details