Gene Transcripts

Gene Symbol
Slc16a10
Entrez Gene ID
170566
Gene Name
solute carrier family 16 (aromatic amino acid transporter), member 10
Gene Aliases
Tat1
Chromosome Location
Chr. 20: 45138433 - 45260158 on Build Rnor_6.0
UniGene ID
Rn.21992
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138831.1 NP_620186.1 4 1153
XM_008772974.1 XP_008771196.1 4 1192
GenBank mRNA AB047324.1 - 4 1153

View Full Product Details