Gene Transcripts

Gene Symbol
Zscan12
Entrez Gene ID
266716
Gene Name
zinc finger and SCAN domain containing 12
Gene Aliases
Zfp96
Chromosome Location
Chr. 17: 45279171 - 45285073 on Build Rnor_6.0
UniGene ID
Rn.44961
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002728485.4 XP_002728531.1 3 1626
GenBank mRNA BC161917.1 - 3 1341

View Full Product Details