Gene Transcripts

Gene Symbol
Ache
Entrez Gene ID
83817
Gene Name
acetylcholinesterase
Gene Aliases
Chromosome Location
Chr. 12: 22472358 - 22477052 on Build Rnor_6.0
UniGene ID
Rn.105879
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172009.1 NP_742006.1 1 273
GenBank mRNA BC094521.1 - 368
S50879.1 - 1 273

View Full Product Details