Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136062.2 | NP_001129534.1 | 4 | 294 | |
NM_001276285.1 | NP_001263214.1 | 4 | 306 | ||
NM_007933.3 | NP_031959.1 | 4 | 406 | ||
GenBank mRNA | AK002485.1 | - | 4 | 266 | |
BC013460.1 | - | 3 | 278 | 4217678 | |
BY115250.1 | - | 4 | 279 | ||
BY784892.1 | - | 4 | 297 | ||
CT010193.1 | - | 3 | 200 | ||
M20745.1 | - | 1 | 26 | ||
X57747.1 | - | 4 | 257 | ||
X62667.1 | - | 3 | 250 |