Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007997.1 | NP_032023.1 | 7, 8 | 816 | |
GenBank mRNA | AK134068.1 | - | 7, 8 | 807 | |
BC057146.1 | - | 7, 8 | 750 | ||
BC071268.1 | - | 7, 8 | 816 | 6517167 | |
D49920.1 | - | 7, 8 | 816 |