Gene Transcripts
- Gene Symbol
- Onecut1
- Entrez Gene ID
- 15379
- Gene Name
- one cut domain, family member 1
- Gene Aliases
- D9Ertd423e, HNF-6, Hfh12, Hnf6, Hnf6b, OC-1, Oc1
- Chromosome Location
- Chr. 9: 74861921 - 74889648 on Build GRCm38
- UniGene ID
- Mm.303355
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U95945.1 |
- |
2 |
1490 |
|
View Full Product Details