Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009621.4 | NP_033751.3 | 8 | 2601 | |
GenBank mRNA | AK156830.1 | - | 8 | 2600 | |
AK161409.1 | - | 8 | 1703 | ||
AK163622.1 | - | 8 | 2600 | ||
BC003269.1 | - | 8 | 2586 | ||
BC040382.1 | - | 8 | 2585 | 3156736 | |
BC050834.1 | - | 8 | 2585 | 6505494 | |
D67076.1 | - | 8 | 2585 |