Gene Transcripts
- Gene Symbol
- LHX6
- Entrez Gene ID
- 26468
- Gene Name
- LIM homeobox 6
- Gene Aliases
- LHX6.1
- Chromosome Location
- Chr. 9: 122185626 - 122228893 on Build GRCh38
- UniGene ID
- Hs.103137
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001242334.1 |
NP_001229263.1
|
8, 9 |
1188 |
|
NM_001242335.1 |
NP_001229264.1
|
6, 7 |
1397 |
|
NM_014368.4 |
NP_055183.2
|
9, 10 |
1238 |
|
GenBank mRNA |
AB031041.1 |
- |
8, 9 |
1085 |
|
AK126982.1 |
- |
6, 7 |
1397 |
|
AK289827.1 |
- |
8, 9 |
1188 |
|
AK299709.1 |
- |
9, 10 |
1272 |
|
AK313808.1 |
- |
8, 9 |
1128 |
|
AL136570.1 |
- |
9, 10 |
1131 |
|
BC103937.1 |
- |
9, 10 |
1097 |
40008210 |
BC103938.1 |
- |
9, 10 |
1094 |
40008211 |
View Full Product Details