Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159543.1 | NP_001153015.1 | 7, 8 | 911 | |
NM_010074.3 | NP_034204.1 | 7, 8 | 911 | ||
GenBank mRNA | AK040409.1 | - | 6, 7 | 617 | |
AK085370.1 | - | 7, 8 | 494 | ||
AK153939.1 | - | 7, 8 | 525 | ||
AK163085.1 | - | 7, 8 | 916 | ||
AK164775.1 | - | 7, 8 | 494 | ||
BC022183.1 | - | 7, 8 | 773 | 3982742 | |
X58384.1 | - | 7, 8 | 588 |