Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289898.1 | NP_001276827.1 | 4 | 368 | |
NM_001289899.1 | NP_001276828.1 | 4 | 840 | ||
NM_013767.6 | NP_038795.3 | 4 | 372 | ||
GenBank mRNA | AB028241.1 | - | 4 | 318 | |
AB028736.2 | - | 4 | 318 | ||
AK158709.1 | - | 4 | 841 | ||
BC026127.1 | - | 4 | 321 | 4010696 |