Gene Transcripts

Gene Symbol
Ltbp1
Entrez Gene ID
268977
Gene Name
latent transforming growth factor beta binding protein 1
Gene Aliases
9430031G15Rik, 9830146M04, Ltbp-1, Tgfb, b2b1000Clo
Chromosome Location
Chr. 17: 75005529 - 75392967 on Build GRCm38
UniGene ID
Mm.269747
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019919.3 NP_064303.2 4, 5 1373
GenBank mRNA AF022889.1 - 4, 5 1338
CV559283.1 - 3, 4 508

View Full Product Details