Gene Transcripts
- Gene Symbol
- Cx3cl1
- Entrez Gene ID
- 20312
- Gene Name
- chemokine (C-X3-C motif) ligand 1
- Gene Aliases
- AB030188, ABCD-3, AI848747, CX3C, Cxc3, D8Bwg0439e, Scyd1
- Chromosome Location
- Chr. 8: 94772180 - 94782427 on Build GRCm38
- UniGene ID
- Mm.103711
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB030188.1 |
- |
1 |
174 |
|
BC057891.1 |
- |
1 |
156 |
|
View Full Product Details